Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC-MS Analysis of Spice Cannabinoids and Metabolites using Titan™ C18
UHPLC-MS Analysis of Spice Cannabinoids and Metabolites using Titan™ C18
Conditions
column Titan C18 10 cm x 2.1 mm, 1.9 µm particles (577124-U)
column temp. 35 °C
mobile phase [A] 0.1% formic acid in 95:5, water:acetonitrile; [B] 0.1% formic acid in 5:95, water:acetonitrile;
gradient 45% B held for 4.5 min; to 80% B in 1.5 min; 80% B held for 4 min
flow rate 0.5 mL/min
pressure 7208 psi (497 bar)
sample 14 Spice cannabinoids 300 ng/mL in 80:20, water:methanol
injection 2 µL
detector MS-ESI+,100 - 1000 m/z scan, XIC
Description
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Forensics and Toxicology
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC