Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Determination of Water Content in 1-Iodine propane, Propyl Iodide Using Karl Fischer Titration
Product Group
Halogenated hydrocarbons
General Information concerning the product group
Halogenated hydrocarbons
Halogenated hydrocarbons can in most cases be titrated by the Karl Fischer method without any problems as is the case for hydrocarbons. No interferences are to be expected from the chemical structure. To overcome solubility problems of unpolar or weakly polar substances, the addition of a solubiliser to the solvent is necessary. In the case of long-chain and cyclic hydrocarbons, long-chain alcohols (e.g. decyl alcohol) or chloroform are thus recommended. Toluene, xylene or chloroform improve the solubility of aromatic compounds. Recommended methods are both the volumetric titration with one or two component reagents, as well as the coulometric analysis. The latter is predominantly applied for low water concentrations (< 0.1 %). If chlorinated hydrocarbons contain free chlorine it is necessary to reduce it prior to KF analysis. This is best achieved through attachment to unsaturated compounds, e.g. through conversion with tetradecene.
Special Information concerning the sample and the methods
Water determination can be carried out without problems according to standard KF titration methods.
Titration one component system
Reagents:
Titrant
188002 Aquastar™ - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
50 mL 188008 Aquastar™ - CombiSolvent - Methanol-free solvent for volumetric Karl Fischer titration with one component reagents
or
50 mL 188009 Aquastar™ - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max 0.01 % water
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
10 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started.
Titration two component system
Reagents:
Titrant
188011 Aquastar™ - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
50 mL 188015 Aquastar™ - Solvent - Solvent for volumetric titration with two component reagents
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
10 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started.
Coulometry with diaphragm
Reagents:
Catholyte
5 mL 109255 Aquastar™ - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
Anolyte
100 mL 109255 Aquastar™ - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
Titration Parameters:
Default coulometer settings for cell with diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample size:
1 - 2 mL
Procedure:
The Karl-Fischer reagent is placed into the cathode and anode compartment of the titration cell with diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started.
Coulometry without diaphragm
Reagents:
Working medium
100 mL 109257 Aquastar™ - CombiCoulomat fritless - Coulometric Karl Fischer reagent for cells with or without diaphragm
Titration Parameters:
Default coulometer settings for cell without diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion: drift < 10 µg/min
Sample size:
1 - 2 mL
Procedure:
The Karl-Fischer reagent is placed into the titration cell without diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started.