Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Determination of Water Content in 2-Bromobenzaldehyde Using Karl Fischer Titration
Product Group
Aldehydes
General Information concerning the product group
Aldehydes
During water determination after Karl Fischer (KF) aldehydes and ketones cause side reactions with the normally utilised methanol. Both side reactions, i.e. the formation of acetals and ketals, as well as bisulphite addition, can often be recognised by the dragging end points and lead to wrong results. During acetal or ketal formation water is released which simulates water contents that are too high. Bisulphite addition, however, consumes water, leading to results that are too low. The extent of the interferences depends on the reactivity of the substances as well as the reactivity of the utilised KF solutions. Aldehydes are more reactive than ketones. Through utilisation of specific KF reagents for aldehydes & ketones it is possible to largely suppress the side reactions. The coulometric method is not recommended.
Special Information concerning the sample and the methods
Aromatic aldehydes are less reactive than aliphatic ones. However, with 2-Bromobenzaldehyde the bromine substituent leads to an increase of side reactions. Here the formation of acetal with methanol and bisulphite addition is strongly characteristic and requires KF titration with specific reagents for aldehydes & ketones. Only small samples can be analysed.
Titration one component system
Reagents:
Titrant
188006 Aquastar™ - CombiTitrant 5 Keto - One component reagent for volumetric Karl Fischer titration of aldehydes & ketones, 1 mL = approx. 5 mg water
Solvent
30 mL 188007 Aquastar™ - CombiSolvent Keto - Solvent for volumetric Karl Fischer titration with one component reagents for aldehydes & ketones
Titration Parameters:
Titration settings for methanol-free solvents:
I(pol) = 20 µA, U(EP) = 100 - 250 mV
Stop criterion for fast end point: drift < 30 µL/min
Sample size:
2 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started. Titration is carried out to fast end point.