Determination of Water Content in Antibiotics Using Karl Fischer Titration
Product Group
Pharmaceuticals
General Information concerning the product group
Pharmaceuticals
Pharmaceutical products are often characterized by complex formulations. Difficulties observed during Karl Fischer determination are often caused by the limited solubility. In some cases side reactions have to be considered. In dependance of composition and properties of the formulations, various measures are necessary for an undisturbed Karl Fischer determination. In pharmaceutical guidelines (USP, Ph Eur, DAB) the Karl Fischer titration is described as common method for water determination. For some substances special procedures can be found. The determination of mass loss as method for water determination is not recommended.
Special Information concerning the sample and the methods
Most types of antibiotics are easily soluble in alcoholic media. Side reactions with the KF reagent only occur in rare cases. Water determination can be carried out according to volumetric standard methods of the one or two component titration. In the case of solubility problems the presence of formamide and / or titration under warm conditions (50 °C) accelerates the release of water.
Titration one component system
Reagents:
Titrant
188005 Aquastar® - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
or
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
50 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
or
30 mL / 20 mL 188009 / 109684 Aquastar® - CombiMethanol / Formamide - solvent mixture for one component titration
Titration Parameters:
Extraction time: 120 sec.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0.2 - 2 g (depending on the water content)
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the powdered sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete dissolution of the sample or rather full extraction of the water a stirring time of 120 seconds is recommended. In the case of hardly soluble samples the titration cell is heated to 50 °C.
Titration two component system
Reagents:
Titrant
188010 Aquastar® - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
or
188011 Aquastar® - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
50 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
or
30 mL / 20 mL 188015 / 109684 Aquastar® - Solvent / Formamide - solvent mixture for two component titration
Titration Parameters:
Extraction time: 120 sec.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0.2 - 2 g (depending on the water content)
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the powdered sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete dissolution of the sample or rather full extraction of the water a stirring time of 120 seconds is recommended. In the case of hardly soluble samples the titration cell is heated to 50 °C.