Determination of Water Content in Chlorotriphenylmethane, Triphenyl chloromethane Using Karl Fischer Titration
Product Group
Halogenated hydrocarbons
General Information concerning the product group
Halogenated hydrocarbons
Halogenated hydrocarbons can in most cases be titrated by the Karl Fischer method without any problems as is the case for hydrocarbons. No interferences are to be expected from the chemical structure. To overcome solubility problems of unpolar or weakly polar substances, the addition of a solubiliser to the solvent is necessary. In the case of long-chain and cyclic hydrocarbons, long-chain alcohols (e.g. decyl alcohol) or chloroform are thus recommended. Toluene, xylene or chloroform improve the solubility of aromatic compounds. Recommended methods are both the volumetric titration with one or two component reagents, as well as the coulometric analysis. The latter is predominantly applied for low water concentrations (< 0.1 %). If chlorinated hydrocarbons contain free chlorine it is necessary to reduce it prior to KF analysis. This is best achieved through attachment to unsaturated compounds, e.g. through conversion with tetradecene.
Special Information concerning the sample and the methods
This substance is hardly soluble in methanol. Titration should thus be carried out in the presence of toluene or long-chain alcohols as solubilisers. Alternatively specific KF solvents for non-polar substances may be used.
Titration one component system
Reagents:
Titrant
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
50 mL 188020 Aquastar® - CombiSolvent oils - Solvent for volumetric Karl Fischer titration with one component reagents for oils
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
2 g
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started.
Titration two component system
Reagents:
Titrant
188011 Aquastar® - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
50 mL 188016 Aquastar® - Solvent oils & fats - Solvent for volumetric Karl Fischer titration with two component reagents for oils & fats
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
2 g
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started.