Determination of Water Content in Hair spray Using Karl Fischer Titration
Product Group
Cosmetics
General Information concerning the product group
Cosmetics
Cosmetics on an aqueous and alcoholic basis can be titrated without problems according to the Karl Fischer (KF) method. Due to the often high water concentration of the emulsions small samples are required. The accuracy of analysis is determined through exact sampling, weighing and adding. The coulometric method is unsuitable for most emulsions. Fatty and oily preparations are insoluble in methanolic KF solvents and require the addition of a solubiliser, or respectively titration with specific KF solvents for oils and fats.
Special Information concerning the sample and the methods
Hair spray is easily soluble in methanolic KF solvents and can be titrated according to volumetric standard methods with one or two component reagents. Due to the relatively high water content the coulometric method is not recommended.
Titration one component system
Reagents:
Titrant
188005 Aquastar® - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
Solvent
50 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
or
50 mL 188008 Aquastar® - CombiSolvent - Methanol-free solvent for volumetric Karl Fischer titration with one component reagents
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0.1 - 0.3 mL
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added with a 1 mL syringe with hollow needle or microlitre syringe (exact determination through weighing of syringe before and after injection) and the titration is started.
Titration two component system
Reagents:
Titrant
188010 Aquastar® - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
Solvent
50 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0.1 - 0.3 mL
Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added with a 1 mL syringe with hollow needle or microlitre syringe (exact determination through weighing of syringe before and after injection) and the titration is started.