Determination of Water Content in Potato chips Using Karl Fischer Titration
Product Group
Foodstuffs
General Information concerning the product group
Foodstuffs
Foodstuffs encompass the most diverse products. Depending on whether carbohydrate-rich, fatty or proteinrich substances are under investigation, different working techniques are favourable. Complex matrices that dissolve slowly, or where the water can only be slowly extracted, necessitate the use of solubilisers. In addition, titration under heating and the application of a homogeniser to accelerate water release are expedient.
Special Information concerning the sample and the methods
The extraction of water in potato chips is sluggish. The presence of formamide during the direct volumetric determination increases the extraction of water. Alternatively, titration may be carried out under warm conditions (50 °C). The use of a homogeniser is also favourable. The coulometric method can only be applied in combination with KF oven technology. For the release of water temperature of 160 °C is recommended.
Titration one component system
Reagents:
Titrant
188005 Aquastar® - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
or
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
30 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
30 mL 109684 Formamide - null
Titration Parameters:
Extraktiontime: 180 sec
Temperatur: 50°C
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
0,15 - 0,2 g (crushed sample; crushing potato chips in the closed bag)