Determination of Water Content in Triethanolamine Using Karl Fischer Titration
Product Group
Alcohols, Amines
General Information concerning the product group
Alcohols
Water determination after Karl Fischer poses no problems for most alcohols. Alcohols are easily soluble in the common Karl Fischer working media. Only alcohols with very long chains require the addition of solubilisers. Side reactions are not to be expected. Suitable methods are both the volumetric titration with one or two component reagents as well as the coulometric determination. The latter method is predominantly applied for low water concentrations (< 0.1%).
Amines
Weakly alkaline amines pose no problems in the water determination after Karl Fischer and can be titrated according to standard methods. For strongly alkaline amines the buffer capacity of the Karl Fischer solution is not sufficient. A shift of pH into the alkaline range leads to a side reaction of the iodine. This can be recognised by the dragging end points, or a continuous consumption of iodine solution, respectively. If turn-off criteria are reached nevertheless, the result is too high. This side reaction, caused by the disproportionation of iodine molecules above pH 8, can be avoided through additional buffering of the working medium with a weak acid. Aquastar® buffer solution for strong bases, benzoic or salicylic acid are the most suitable because, unlike acetic acid, they do not show any tendency for esterification in the Karl Fischer solution. The acids are added to the solvent prior to titration and can even be employed in excess. Some amines react with the Karl Fischer solution. Measures to suppress or overcome this side reaction are, for example, titration in methanol-free working media, utilisation of small sample sizes, as well as titration to a fast end point.
Special Information concerning the sample and the methods
Triethanolamine possesses strong alkaline properties. Titration with two component reagents is more suitable due to the better buffering of the KF solvent. Through addition of benzoic or salicylic acid the amount of substance that can be determined in the KF solvent increases. The hygroscopic properties of triethanolamine require careful sample handling.
Titration one component system
Reagents:
Titrant
188005 Aquastar® - CombiTitrant 5 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
or
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
50 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
5 g 100635 salicylic acid - as buffer substance
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion for fast titration: drift < 30 µL/min
Sample size:
3 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started. Titration is carried out to fast end point.
Titration two component system
Reagents:
Titrant
188010 Aquastar® - Titrant 5 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 5 mg water
or
188011 Aquastar® - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
50 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
and
5 g 100635 salicylic acid - as buffer substance
Titration Parameters:
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion for fast titration: drift < 30 µL/min
Sample size:
3 mL
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added with a syringe (exact sample weight determination by weighing of syringe before and after injection) or volumetric pipette and the titration is started. Titration is carried out to fast end point.
Coulometry with diaphragm
Reagents:
Catholyte
5 mL 109255 Aquastar® - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
Anolyte
100 mL 109255 Aquastar® - CombiCoulomat frit - Coulometric Karl Fischer reagent for cells with diaphragm
and
20 g 100136 benzoic acid - as buffer substance
Titration Parameters:
Default coulometer settings for cell with diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion for fast titration: drift < 15 µg/min
Sample size:
0.5 - 1 mL
Procedure:
The Karl-Fischer reagent is placed into the cathode and anode compartment of the titration cell with diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started. Titration is carried out to fast end point.
Coulometry without diaphragm
Reagents:
Working medium
100 mL 109257 Aquastar® - CombiCoulomat fritless - Coulometric Karl Fischer reagent for cells with or without diaphragm
and
20 g 100136 benzoic acid - as buffer substance
Titration Parameters:
Default coulometer settings for cell without diaphragm:
For end point indication, e.g.:
I(pol) = 5 - 10 µA, U(EP) = 50 - 100 mV
Stop criterion for fast titration: drift < 15 µg/min
Sample size:
0.5 - 1 mL
Procedure:
The Karl-Fischer reagent is placed into the titration cell without diaphragm. The coulometer is started and the solvent is titrated dry. After preliminary titration and stabilisation of drift the sample is injected into the titration cell with a syringe (exact sample weight determination by weighing of syringe before and after injection) and the water determination is started. Titration is carried out to fast end point.