Analytical Method: Aluminium (total) in ores
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 100594  Spectroquant® Aluminium Cell Test
Cat.No. 114825  Spectroquant® Aluminium Test
Cat.No. 106308  di-Sodium tetraborate decahydrate for analysis
Cat.No. 100317  Hydrochloric acid 37 % for analsis
Cat.No. 116754  Water for analsis
Sample preparation:
Mix about 100 mg of very finely powdered material with 1.5 g of di-sodium tetraborate in a platinum crucible and heated at 900 °C for 30 minutes in the muffle furnace. After cooling, dissolve the content of the crucible in an uncovered glass beaker with 2 ml of hydrochloric acid 37 % and 50 ml water for analysis. When fully dissolved fill up to volume in a 1000-ml volumetric flask with water for analysis and mix well.
Determine with the above mentioned test kits.
Aluminium (total) content in %  =   analysis value in mg/l Al x 100
weight of sample in mg
Related Links