Analytical Method: Aluminium (total) in steels (Others)
Instruments and Reagents:
Cat.No. 173016 Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017 Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018 Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748 Spectroquant® Photometer NOVA 30 or
Cat.No. 109751 Spectroquant® Photometer NOVA 60 or
Cat.No. 173632 Spectroquant® Colorimeter Move 100
Cat.No. 114946 Rectangular cells 10 mm and/or
Cat.No. 114947 Rectangular cells 20 mm and/or
Cat.No. 114944 Rectangular cells 50 mm
Cat.No. 100594 Spectroquant® Aluminium Cell Test
Cat.No. 114825 Spectroquant® Aluminium Test
Cat.No. 100317 Hydrochloric acid 37 % for analsis
Cat.No. 100456 Nitric acid 65 % for analysis
Cat.No. 116754 Water for analysis
Sample preparation:
In a glass beaker dissolve about 100 mg of sample in 3 ml hydrochloric acid 37 % and 2 ml nitric acid 65 %, on the steambath. When completely dissolved evaporate the acid until only a damp crystalline residue remains. Dissolve this residue in 1 ml hydrochloric acid 37 % and 20 ml water for analysis, transfer to a volumetric flask and fill up to 100 ml with water for analysis.
Determine with the above mentioned test kits.
Aluminium (total) content in %  =   analysis value in mg/l Al x 10
weight of sample in mg
Related Links