Analytical Method: Ammonium in disposal drainage water
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm and/or 
Cat.No. 114558  Spectroquant® Ammonium Cell Test
Cat.No. 114544  Spectroquant® Ammonium Cell Test*
Cat.No. 114559  Spectroquant® Ammonium Cell Test
Cat.No. 114752  Spectroquant® Ammonium Test
Cat.No. 100683  Spectroquant® Ammonium Test
*not compatible with Move 100
Sample preparation:
Dilution 1:100 for influx and 1:10 for efflux.
Determine with the above mentioned test kits.
Ammonium content in mg/l NH4 = analysis value in mg/l NH4 x 100 or x 10 depending on the dilution.
Related Links