Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Ammonium in soils
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114739  Spectroquant® Ammonium Cell Test
Cat.No. 114558  Spectroquant® Ammonium Cell Test
Cat.No. 114544  Spectroquant® Ammonium Cell Test*
Cat.No. 114559  Spectroquant® Ammonium Cell Test
Cat.No. 114752  Spectroquant® Ammonium Test
Cat.No. 100683  Spectroquant® Ammonium Test
Cat.No. 102382  Calcium chloride dihydrate for analysis
Cat.No. 116754  Water for analysis
Charcoal activated for soil tests
Folded filter
*not compatible with Move 100
Calcium chloride solution 0.025 mol/l:
Dissolve 3.68 g calcium chloride dihydrate for analysis with 1 l of water for analysis.
Sample preparation:
In a glass bottle mix 50 g of naturally moist sample, free from coarse stones, with 100 ml of the calcium chloride solution 0.025 mol/l. Add a spatula - tipfull of charcoal activated for soil tests and shake the closed bottle in a shaking machine for 1 hour (alternative stir in a beaker). Let the suspension settle and filter it through a folded filter.
For determination of the water - content dry a similar sample to constant weight in the drying kiln at 105 °C. A drying of the sample before determination is not advisable because of the fast change in the nitrogen-forms. The analysis should take place very quickly. Storing the sample in plastic bags at room temperature changes the analysis values after a short time already.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Ammonium content in mg/kg NH4 = analysis value in mg/l NH4 x 2
Materials
Related Links
Categories