Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Acid Capacity
Reagents:
Cat.No. 109141  Sodium hydroxide solution 0,1 mol/l
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
A sodium hydroxide solution 0.1 mol/l (corresponds to 100 mmol/l) can be used.
Further investigational concentrations may be prepared from this stock solution by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one week.
For use with test kits:
Measuring range
in mmol/l
101758
Spectroquant®Acid Capacity Cell Test to pH 4.3
0.20-8.00