Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Free Chlorine (sodium hypochlorite solution)
Preparation of a standard solution for free chlorine
Reagents:
Cat.No. 100316  Hydrochloric acid 25 % for analysis
Cat.No. 105614  Sodium hypochlorite solution (6 - 14 % active chlorine)
Cat.No. 109147  Sodium thiosulfate solution 0.1 mol/l 0.1 N solution
Cat.No. 105043  Potassium iodide for analysis
Cat.No. 105445  Zinc iodide starch solution for analysis
Cat.No. 116754  Water for analysis
Preparation of a stock solution of free chlorine:
First prepare a 1:10 dilution using a sodium hypochlorite solution containing approximately 6 - 14 % of active chlorine. For this pipette 10 ml of sodium hypochlorite solution into a calibrated or conformity-checked 100-ml volumetric flask and then make up to the mark with water for analysis.
Precise assay of the stock solution for free chlorine:
Pipette 10.0 ml of the stock solution into a 250-ml ground-glass-stoppered conical flask containing 60 ml of water for analysis. Subsequently add to this solution 5 ml of hydrochloric acid 25 %. And 3 g of potassium iodide. Close the conical flask with the ground-glass stopper, mix thoroughly, and leave to stand for 5 min. Titrate the eliminated iodine with sodium thiosulfate solution 0.1 mol/l until a weakly yellow color emerges. Add 2 ml of zinc iodide starch solution and titrate from blue to colorless.
Calculation:
1 ml sodium thiosulfate solution = 3.55 mg free chlorine
Further investigational concentrations may be prepared from the stock solution prepared according to the procedure described above by diluting accordingly.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for approximately one week. The diluted investigational solutions are stable for approximately 2 hours.
For use with test kits:
Measuring range
in mg/l Cl2
117925
MQuantTM Chlorine Test (free chlorine)
4 - 120
117924
MQuantTM Chlorine Test (free chlorine)
25 - 500
114434
MColortestTM Chlorine Test (free chlorine)
0.01 - 0.3
111174
MColortestTM Chlorine and pH Test (free + total chlorine)
0.1 - 1.5
111100
MColortestTM Chlorine and pH Test (free + total chlorine)
refill pack for 111174
111113
111114
MColortestTM Chlorine Test Reagent Cl2-1 
MColortestTM Chlorine Test Reagent Cl2-2
111160
MColortestTM Chlorine and pH Test (free chlorine)
0.1 - 1.5
114670
MColortestTM Chlorine Test (free chlorine)
0.1 - 2
114978
MColortestTM Chlorine Test (free chlorine, liquid)
0.1 - 2
114979
MColortestTM Chlorine Test (free chlorine, liquid)
refill pack for 114978 
1.14801
MColortestTM Chlorine Test (free + total chlorine, liquid)
0.1 - 2
114803
MColortestTM Chlorine Test (free + total chlorine, liquid)
refill pack for 114801
114976
MColortestTM Chlorine Test (free chlorine)
0.25 - 15
114977
MColortestTM Chlorine Test (free chlorine)
refill pack for 114976
114826
MColortestTM Chlorine Test (free + total chlorine)
0.25 - 15
118326
MColortestTM Chlorine Test (free + total chlorine)
refill pack for 114826
116896
Reflectoquant® Chlorine Test (free + total chlorine)
0.5 - 10.0
100595
Spectroquant® Chlorine Cell Test (free chlorine)
0.03 - 6.00
100598
Spectroquant® Chlorine Test (free chlorine)
0.010 - 6.00
100086
100087
Spectroquant® Chlorine Reagents Cl2-1
Spectroquant® Chlorine Reagents Cl2-2
0.010 - 6.00
119254
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 10-ml-samples (free chlorine)
0 - 2.00
119256
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 25-ml-samples (free chlorine)
0 - 10.00
100597
Spectroquant® Chlorine Cell Test (free + total chlorine)
0.03 - 6.00
100599
Spectroquant® Chlorine Test (free + total chlorine)
0.010 - 6.00