Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Oxygen
Preparation of a standard solution (investigational concentration):
Prepare the standard solution by bubbling air through tap water in a wash bottle for approx. 1 hour.
Then measure the water temperature and immediately measure the oxygen concentration.
Stability:
The standard solution is not stable and must be used immediately.
Evaluation:
Read off the theoretical oxygen concentration (specified value) of the prepared standard solution from the table "Oxygen saturation concentration of water" acc. to Oehme and Schuler, 1983 (see page 2).
The oxygen concentration measured using a test kit should not deviate from the value read off from the oxygen saturation table by more than + 0.1 %.
For use with test kits:
Measuring range
in mg/l O2
114662
MColortestTM Oxygen Test
1 -12
111107
MColortestTM Oxygen Test
0.1 - 10 with one full pipette (graduation 0.1)
114694
Spectroquant® Oxygen Cell Test
0.5 - 12.0
Table for calculating oxygen saturation
Materials
Related Links
Categories