Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Analytical Quality Assurance, Standard for Volatile Organic Acids
Reagents:
Cat.No. 106267  Sodium acetate trihydrate for analysis
Cat.No. 116754  Water for analysis
Preparation of a standard solution:
Dissolve 3.40 g of sodium acetate trihydrate with distilled water in a calibrated or conformity-checked 1000-ml volumetric flask and make up to the mark with distilled water. The stock solution prepared according to this procedure has a concentration of 1500 mg/l acetic acid.
Stability:
When stored in a cool place (refrigerator), the stock solution remains stable for one week.
For use with test kits:
Measuring range
in mg/l HOAc
101749
Spectroquant® Volatile Organic Acids Cell Test
50 - 3000
101809
Spectroquant® Volatile Organic Acids Test
50 - 3000