Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Aniline in wastewater
Reagents and instruments:
Cat.No. 106549 Sodium nitrite for analysis
Cat.No. 100103 Amidosulfuric acid for analysis
Cat.No. 106237 N-(1-Naphtyl)-ethylendiamine dihydrochloride for analyis
Cat.No. 116754 Water for analysis
Cat.No. 100317 Hydrochlorid acid 37 % for analysis
Cat.No. 114724 Empty cells 16 mm with screw caps
Folded filter
Membrane filter 0.45 μm
Waterbath
Reagent 1:
Dissolve 40 g sodium nitrite in 60 ml water
Reagent 2:
Dissolve 15 g amidosulfuric acid in 85 ml water.
Reagent 3:
N-(1-naphthyl)ethylenediamine dihydrochloride (NEDDC)
Measurement range:
0.5 - 8.0 mg/l Aniline at cell length 16 mm and 550 nm
Sample preparation:
The sample solution must be clear; if not, filter through a folded filter or a membrane filter.
Analysis:
Adjust the clear sample to pH 2-3 with hydrochloric acid and place 5.0 ml in an empty cell. Close off the cell and cool in an ice bath for 3 minutes. Add 0.10 ml reagent 1 and mix and 2.0 ml of reagent 2 and shake vigorously for 30 s. Add approx. 10 mg of reagent 3 and mix. Close off the cell and heat in a water bath for 5 minutes at 60 °C. Allow to cool at room temperature for 10 minutes, dry the cell and measure at 550 nm (in SQ 118 against distilled water) in a round cell.
Parameter list for programming the photometer Spectroquant® NOVA 60 und NOVA 400
Method number ----
Method-designation Aniline
Wavelength 550 nm
Citation Aniline
Units mg/l
Resolution 0.1
Zero point (E0) 0.000
Slope (E/C) 0.333
Meas. range begin 0.5
Meas. range end 8.0
Reference cell 16 mm round
Calculation when measuring the absorbance value using the above NOVA photometers without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/l
[A sample * 1 / 0.333]
1.000 3.00
Parameter list for programming the photometer SQ 118
01 Method Aniline
02 Cat.No. -------
03 Measuring range 0.5 - 10.0
04 Units mg/l
05 Reaction time 5 10 min
06 Cell 16 mm round
07 Wavelength 550 nm
08 Calibration Factor
09 Evaluation Linear
10 Factor 3.00
Calculation when measuring the absorbance value using the photometer SQ118 without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/l
[A sample * 3.00]
1.000 3.00
Materials
Related Links
Categories