Analytical Method: Boron in aluminium and aluminium alloys
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114839  Spectroquant® Boron Test
Cat.No. 100456  Nitric acid 65 % for analysis
Cat.No. 105591  Sodium hydroxide solution 27 % for analysis
Cat.No. 106549  Sodium nitrite for analysis
Cat.No. 116754  Water for analysis
Sample preparation:
In a PTFE-melting-pot mix about 500 mg of the sample with 10 ml sodium hydroxide solution 27 % and dissolve under warming. Add 50 ml water for analysis and 15 ml nitric acid 65 % and boil the mixture to clearness. Possibly precipitated manganese dioxide can be reduced and dissolved by adding some sodium nitrite. Put the clear solution into a 250-ml volumetric flask, fill up to volume with water for analysis and mix well.
Determine with the above mentioned test kit.
Boron content in % =   analysis value in mg/l B x 25
weight of sample in mg
Related Links