Analytical Method: Calcium, low measuring range
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114815  Spectroquant® Calcium Test
Cat.No. 116754  Water for analysis
Measuring range:
1.0 - 15.0 mg/l calcium in a 10-mm cell at 550 nm.
To 1 ml sample add 10 ml of reagent Ca-1 and mix. Add 8 drops of reagent Ca-2 and Ca-3 and mix.
For the blank reading treat 1 ml of water for analysis in the same way as the sample.
Determine by means of photometer; the method is already programmed in the instruments.
Related Links