Analytical Method: Chloride in red and white wines
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114730  Spectroquant® Chloride Cell Test
Cat.No. 114897  Spectroquant® Chloride Test
Cat.No. 116754  Water for analysis
Clarocarbon® G activated charcoal
Folded filter
Sample preparation:
Mix 100 ml of the sample of wine with 5 g Clarocarbon® G activated charcoal and stir for 10 minutes. Use an analogous mixture of 100 ml water for analysis and 5 g activated charcoal as blank. Filter the charcoal through a folded filter. The solution must be clear and colorless.
Determine with the above mentioned test kits.
Chloride content in mg/l Cl = analysis value in mg/l Cl
Related Links