Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Copper (total) in electroplating baths
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114553  Spectroquant® Copper Cell Test
Cat.No. 114767  Spectroquant® Copper Test
Cat.No. 100317  Hydrochloric acid 37 % for anaylsis
Cat.No. 104944  Potassium chlorate for analysis
Cat.No. 116754  Water for analysis
Hints:
The fact that complex-compounds will be present means that an appropriate pretreatment is necessary.
Sample preparation:
In a glass beaker place 10 ml of the bath fluid with 10 ml of hydrochloric acid 37 % and 3 g of potassium chlorate, mix well and evaporate, on the heating plate, almost to dryness. Dissolve the residue in water for analysis, transfer to a 1000-ml volumetric flask, fill up to volume with water for analysis and mix well.
Analysis:
Determine with the above mentioned test kits
Calculation:
Copper (total) content in mg/l Cu = analysis value in mg/l Cu x 100
Materials
Related Links
Categories