Analytical Method: Determination of the Solution Color of Raw Sugars, Brown Sugars and Colored Syrups at pH 7.0 (According to ICUMSA Method GS1/3-7)
Materials and Reagents:
Cat.No. 1.16754.9010  Water for analysis EMSURE® or distilled water
Cat.No. 1.09060.1000  Hydrochloric acid 0.1 mol/l Titripur®
Cat.No. 1.09141.1000  Sodium hydroxide solution 0.1 mol/l Titripur®
Cat.No. 1.14946.0001  Rectangular cells 10 mm or
Cat.No. 1.14947.0001  Rectangular cells 20 mm or
Cat.No. 1.14944.0001  Rectangular cells 50 mm
Cat.No. 1.73016.0001 Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 1.73017.0001 Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 1.73018.0001 Spectroquant® UV/VIS Spectrophotometer Prove 600
Membrane filters, cellulose nitrate, pore size 0.45 µm, diameter 50 mm
Conical flasks 250 ml
Volumetric flask 500 ml
pH meter
Ultrasonic bath
Refractometer
Method:
Spectrophotometric Color Determination at 420 nm
Measuring range:
Method No. 2548
0 – 50 000 IU7.0
Working instructions:
Sample preparation
Mix the sample thoroughly.
Weigh quantities of sample and water as shown in “Table 1” below into a 250 ml conical flask.
Dissolve the sample by swirling at room temperature.
Adjust the pH of the prepared solution to 7.0 ± 0.1 with Hydrochloric acid 0.1 mol/l Titripur® or, respectively, Sodium hydroxide solution 0.1 mol/l Titripur® as required.
Vacuum-filter the pretreated solution over a membrane filter into a clean and dry conical flask.
Filter the sample solution under vacuum through a membrane filter into a dry conical flask.
Degas the solution for 3 minutes in the ultrasonic bath.
Measure the % RDS (refractometric dry substance) of the solution to an accuracy of ± 0.1 g/100g (according to ICUMSA Method GS4/3-13).
Table 1
Aliquots of sample and water (Water for analysis EMSURE® is recommended)
and recommended cell length
  ICUMSA Color    
Cell length Range Sample Water
[mm] [IU7.0] [g] [g]
  <250 50 + 1 50 + 1
50 250 - 500 30 + 1 70 + 1
  500 - 2000 30 + 1 70 + 1
  500 - 2000 30 + 1 70 + 1
20 2000 - 7000 10 + 1 90 + 1
  7000 - 25000 5.0 + 0.1 95 + 1
  500 - 2000 30 + 1 70 + 1
10 2000 - 7000 10 + 1 90 + 1
  7000 - 25000 5.0 + 0.1 95 + 1
  >25000 5.0 + 0.1 95 + 1
Zero adjustment
For each measurement series, a zero adjustment is required.
It is recommended to use the same cell for zero adjustment and for sample measurement.
The zeroing procedure for the measurement series is automatically prompted by the instrument after entering the %RDS value.
For zero adjustment fill a corresponding rectangular cell with distilled water or water for analysis.
After prompting, place the rectangular cell filled with water in the cell compartment, the zero adjustment is executed automatically.
Confirm the implementation of zero adjustment with OK.
The zero adjustment is valid for the entire measurement series.
Measurement procedure
Select the method (2548, "ICUMSA Color GS1/3-7") from the method list of the Spectroquant® Prove. 
Enter the RDS value in %. Confirm with OK.
Press START to start the measurement mode.
Perform a zero adjustment (procedure see “Zero adjustment”).
Transfer the sample solution into a corresponding cell. Place the cell into the cell compartment. The measurement is performed automatically.
Confirm measurement with OK. The IU7.0 value is shown in the display.
Press START to start the measurement procedure for the next sample. The system does not prompt a repeat of the zero adjustment.
Note:
According to ICUMSA Method GS1/3-7 the method checks automatically if the
measured absorption is below 2.500 A. If the absorption is not in this range, a
pop-up message appears: “Condition not met – Absorbance > 2.5”.
In these cases, the sample preparation needs to be adjusted. Use an appropriate cell length, sample and water weight according to recommendations of Table 1.
The result is calculated according to ICUMSA GS1/3-7(2011) chapter 8.1 and ICUMSA SPS-4 table B (Sucrose; 20°C).
Individual ICUMSA methods or references are available online within 48 h: www.icumsa.org under “Methods book”.
Short measuring procedures on Spectroquant® Prove instruments are described in the Analytical Procedures and Appendices, available online on our website.