Analytical Method: Formaldehyde in cooling lubricants
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114500  Spectroquant® Formaldehyde Cell Test
Cat.No. 114678  Spectroquant® Formaldehyde Test
Cat.No. 100716  Sulfuric acid 25 % for analysis
Cat.No. 105588  Sodium hydroxide solution 10 % for analysis
Cat.No. 109533  MColorpHastTM pH indicator strips pH 5 - 10
Cat.No. 110537  Carrez clarification
Cat.No. 116754  Water for analysis
As samples can be either acid or alkaline, the pH of the ready-to-use emulsion must be adjusted to neutral with acid or alkali prior to determining HCHO. This facilitates precipitation.
Sample preparation:
Adjust the ready-to-use emulsion to pH 6 - 8 with either 25 % sulfuric acid or 10 % sodium hydroxide solution. Place approx. 10 ml of the sample in a 200-ml volumetric flask, add approx. 100 ml water for analysis and mix. Add 2 ml of Carrez solution 1 and 2 consecutively, mixing well after each addition. Make up to the mark with water for analysis and mix well. Allow the precipitate to settle. Filter the supernatant through a paper filter; the filtrate must be clear.
Determine with the above mentioned test kits.
Formaldehyde content in mg/l HCHO = analysis value in mg/l HCHO x 20
Related Links