Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Gold in effluents
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114821  Spectroquant® Gold Test
Sample preparation:
The determination of dissolved mono- and trivalent gold compounds and the differentiation between Au(I) and Au(III), requires no special preparation of the sample.
For the determination of the Au(III)-content, treatment with Reagent Au-1 is omitted. After the usual procedure described in the attached slip, the total Au(III) and Au(I) can be calculated.
Analysis:
Determine with the above mentioned test kit.
Calculation:
Gold content in mg/l Au = analysis value in mg/l Au
Materials
Related Links
Categories