Analytical Method: Iron in hexacyanoferrate solutions
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114549  Spectroquant® Iron Cell Test
Cat.No. 114896  Spectroquant® Iron Cell Test*
Cat.No. 114761  Spectroquant® Iron Test
Cat.No. 100796  Spectroquant® Iron Test
Cat.No. 100456  Nitric acid 65 % for analysis
Cat.No. 116754  Water for analysis
*not compatible with Move 100
Sample preparation:
In a 250-ml glass beaker (wide shape) mix 10 ml of the sample with 2 ml nitric acid 65 % and evaporate. Dissolve the residue in 10 ml water for analysis.
Determine with the above mentioned test kits.
Iron content in mg/l Fe = analysis value in mg/l Fe
Related Links