Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Manganate and Permanganate in pickling baths
Reagents:
Cat.No. 109141 Sodium hydroxide solution 0.1 mol/l
Sample preparation:
Pipette 5 ml of bath solution into a 50-ml standard volumetric flask, fill up to volume with sodium hydroxide solution 0.1 mol/l and mix well. From this solution pipette 1 ml into 100-ml standard volumetric flask and fill up to volume with sodium hydroxide solution 1 mol/l and mix well, too. This solution can be used for determination.
Attention: The determination has to take place as quickly as possible, because the sample changes after dilution.
Analysis:
Determine the K2MnO4 - and the KMnO4 - content by measuring the absorbance at 525 nm and 585 nm in cells of 10-mm cell length against sodium hydroxide solution 0.1 mol/l as a blank in the SQ 118 photometer.
Calculation:
Manganate:
K2MnO4 content in g/l = (A585 x 105.86) - (A525 x 12.39)
Permanganate:
KMnO4 content in g/l = (A585 x 65.83) - (A525 x 16.36)
A= Absorbance at wavelength.
Materials
Related Links
Categories