Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Manganese (total) in ores (Others)
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 100816  Spectroquant® Manganese Cell Test
Cat.No. 114770  Spectroquant® Manganese Test
Cat.No. 101846  Spectroquant® Manganese Test
Cat.No. 101145  Ammonium chloride for analysis
Cat.No. 116754  Water for analysis
Ascorbic acid for analysis
Sample preparation:
In a test tube place about 100 mg of finely powdered sampling material together with 2 g of ammonium chloride for analysis and mix well. Then heat over a flame to complete sublimation (fume cupboard!). In the upper third of the tube a white sublimate will be found, in the midportion, an orange-yellow, whilst the lower zone is clear of siliceous residue.
Add, after cooling, about 1 g of ascorbic acid for analysis and 5 ml of water for analysis and heat for a short period. Then transfer the solution to a 1000-ml standard volumetric flask, fill up to the mark with water for analysis and mix well.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Manganese (total) content in % =   analysis value in mg/l Mn x 100
weight of sample in mg