Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Nickel (total) in effluents, high organic content
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114554  Spectroquant® Nickel Cell Test
Cat.No. 114785  Spectroquant® Nickel Test
Cat.No. 100731  Sulfuric acid 95 - 97 % for analysis
Cat.No. 107209  Hydrogen peroxide 30 % (Perhydrol®) for analysis
Cat.No. 105590  Sodium hydroxide solution 32 % for analysis
Cat.No. 109535  MColorpHastTM Universal Indicator strips pH 0 - 14
Cat.No. 116754  Water for analysis
Sample preparation:
Place 25 ml sample in a 400-ml beaker along with 5 ml sulfuric acid 95 - 97 % and 5 ml Perhydrol®, mix and slowly heat on a heating plate until the sulfuric acid begins to fume. Allow to cool, add 5 ml Perhydrol® and heat again until SO3 is released. Depending on the organic content of the sample, repeat this process up to three times. The fully digested sample should be clear and practically colorless.
Transfer the mixture to a 50-ml volumetric flask. Adjust to pH 4 - 7 with sodium hydroxide solution 32 % (control with Universal Indicator strips pH 0 - 14). Dilute the solution with water for analysis fill up to the mark and mix well.
Analysis:
Determine with the above mentioned test kits
Calculation:
Nickel (total) content in mg/l Ni = analysis value in mg/l Ni x 2