Analytical Method: Nitrate in hops (Photometry)
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114542  Spectroquant® Nitrate Cell Test
Cat.No. 114563  Spectroquant® Nitrate Cell Test*
Cat.No. 114764  Spectroquant® Nitrate Cell Test*
Cat.No. 114773  Spectroquant® Nitrate Test
Cat.No. 109713  Spectroquant® Nitrate Test*
Cat.No. 116754  Water for analysis
*not compatible with Move 100
Sample preparation:
In a 200-ml iodine-number flask place 5 g of hop pellets with 100 ml of water for analysis, close and warm on the waterbath for 60 minutes (about 50 °C). Pour the resulting suspension through a folded filter to remove the undissolved portion.
Dilute the filtrate 1:100 with water for analysis.
Determine with the above mentioned test kits.
Nitrate content in % = analysis value in mg/l NO3 x 0.2
Related Links