Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Nitrite in disposal drainage water
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114547  Spectroquant® Nitrite Cell Test
Cat.No. 114776  Spectroquant® Nitrite Test
Cat.No. 110537  Carrez Clarification
Cat.No. 100716  Sulfuric acid 25 % for analysis
Cat.No. 109533  MColorpHastTM pH-indicator strips pH 0 - 6
Cat.No. 116754  Water for analysis
Sample preparation:
In a 100 ml of standard volumetric flask mix 90 ml of the sample solution (pH 6 - 7) with 2 ml each Carrez solution-1 and -2 and fill up to volume. Filter off the occuring deposit through a folded filter and adjust the clear, light yellow filtrate to pH 3 using sulfuric acid 25 % for analysis (check with pH-indicator strips).
Analysis:
Determine with the above mentioned test kits.
Calculation:
Nitrite content in mg/l NO2 = analysis value in mg/l NO2