Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Nitrogen (total) in fish meal
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114542  Spectroquant® Nitrate Cell Test
Cat.No. 114773  Spectroquant® Nitrate Test
Cat.No. 100731  Sulfuric acid 95 - 97 % for analysis
Cat.No. 107209  Hydrogen peroxide 30 % (Perhydrol®) for analysis
Cat.No. 105590  Sodium hydroxide solution 32 %
Cat.No. 116754  Water for analysis
Sample preparation:
Weigh exactly approximately 1 g sample into a 600-ml beaker, add 5 ml sulfuric acid 95 - 97 % and slowly evaporate on an electric plate until the solution becomes black. Allow to cool and then add Perhydrol® in 1-ml portions. Vigorous reaction accompanied with foaming will take place. Continue heating the mixture until the fume point is reached. If the solution becomes brown or black, add Perhydrol® until it again becomes colorless. In this example, approximately 10 ml Perhydrol® would be required for oxidation of the sample. Allow to cool, dilute with ca. 50 ml water for analysis in a volumetric flask, adjust to pH 5 - 7 with 14 - 15 ml sodium hydroxide solution
32 % using a pH-meter and make up to 1 l with water for analysis. Dilute this solution to 1 l in a volumetric flask.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Nitrogen (total) content in mg/kg N = analysis result in mg/l NO3-N x 1000