Analytical Method: Phosphate in beer
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114543  Spectroquant® Phosphate Cell Test
Cat.No. 114729  Spectroquant® Phosphate Cell Test
Cat.No. 114848  Spectroquant® Phosphate Test
Cat.No. 116754  Water for analysis
Sample preparation:
Degas 100 ml beer in an ultrasonic bath. Dilute the carbon dioxide-free sample 1:199 with water for analysis.
Determine with the above mentioned test kits.
Phosphate content in mg/l PO4 = analysis value in mg/l PO4 x 200
Related Links