Analytical Method: Phosphorus in textile finish solutions
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114543  Spectroquant® Phosphate Cell Test
Cat.No. 114729  Spectroquant® Phosphate Cell Test
Cat.No. 116754  Water for analysis
Sample preparation:
Cleaning solutions contain, beside from pebble brine (highly disperged silicic acid), condensed phosphates and soluble phosphorus acidic ester. In a standard volumetric flask dilute the sample according to the expected
P-content 1:100 to 1:200 with water for analysis and filter it.
Determine with the above mentioned test kits.
Multiply the analysis value with the factor of dilution of the sample The result is equal to phosphorus (total) content in the original sample.
Related Links