Analytical Method: Phosphorus (total) in steels (Others)
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114543  Spectroquant® Phosphate Cell Test
Cat.No. 114729  Spectroquant® Phosphate Cell Test
Cat.No. 114848  Spectroquant® Phosphate Test
Cat.No. 100317  Hydrochlorid acid 37 % for analysis
Cat.No. 100456  Nitric acid 65 % for analysis
Cat.No. 116754  Water for analysis
Sample preparation:
In a well covered glass beaker dissolve about 100 mg of sample in 3 ml hydrochloric acid 37 % and 3 ml nitric acid 65 % on the steambath. When completely dissolved, evaporate the acids, to a well-dried, acid-free, crystalline solid residue.
After taking up into a few ml of water for analysis, transfer the solution to a 1000-ml volumetric flask, fill to the mark with water for analysis and mix well.
Determine with the above mentioned test kits.
Phosphorus (total) content in %  =   analysis value in mg/l P x 5
weight of sample in mg
Related Links