Analytical Method: Silicon (total) in iron oxide powder
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114794  Spectroquant® Silicate (Silicic Acid) Test
Cat.No. 100716  Sulfuric acid 25 % for analysis
Cat.No. 106495  Sodium hydroxide for analysis
Cat.No. 109533  MColorpHastTM pH-indicator strips pH 5 - 10
Cat.No. 116754  Water for analysis
Iron oxide powder with an average size of particles of 0.2 - 0.3 mm is used for the production of ferrite cores. Mineral and oxidic admixtures or contaminations are decisive for the quality of the endproduct.
Sample preparation:
In a nickel melting-pot mix 100 mg of the sample, dried to constant weight at 105 °C, with 8 sodium hydroxide tablets (1.5 g). Directly with the gas burner heat the covered melting-pot to weak red glow for 30 minutes. After cooling dissolve the content of the melting-pot in water for analysis. For that purpose put the melting-pot with the cover into a glass beaker as well as 30 - 50 ml water for analysis warm on a bath of water, until the melting cake has dissolved. Adjust the suspension to a pH-value of 7 - 8 with sulfuric acid 25 %, quantitatively transfer it into a 250-ml standard volumetric flask and fill up to volume with water for analysis. After mixing let the insoluble parts settle and filter the supernatant solution through an ash-free, acid washed filter. A blank reading analysis must be considered, produced with the used reagents according to the procedure above.
Determine with the above mentioned test kit.
Silicon (total) content in %  =   analysis value in mg/l Si x 25
weight of sample in mg
Related Links