Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Vanadium in water (Others)
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 
Cat.No. 104936  Potassium chloride for analysis
Cat.No. 109057  Hydrochloric acid 1 mol/l
Cat.No. 100983  Ethanol for analysis
Cat.No. 116754  Water for analysis
2-Thiophen-carboxylic hydrazide, Aldrich Cat.No. T 3,261-1
Membrane filter 0.45 μm pore diameter
Reagent 1:
Dissolve 6 g 2-thiophen-carboxylic hydrazide in 40 ml ethanol.
Reagent 2:
Place 12.5 ml potassium chloride solution, 0.2 mol/l and 3.25 ml hydrochloric acid, 0.2 mol/l in a 50-ml volumetric flask and make up to the mark with water.
Measurement range:
0.50 - 5.00 mg/l V at cell length 10 mm and 405 nm
Sample preparation:
The sample solution must be clear; if not, filter through a folded filter or membrane filter with 0.45 μm pore diameter.
Analysis:
Mix 10.0 ml of sample solution, 0.25 ml reagent 1, 0.30 ml reagent 2 in a test tube. Measure against water at 405 nm.
Parameter list for programming the photometer Spectroquant® NOVA 60, NOVA 400, Pharo 100, and Pharo 300
Method number ----
Method-designation Vanadium
Wavelength 405 nm
Citation V
Units mg/l
Resolution 0.01
Zero point (E0) 0.000
Slope (E/C) 0.2188
Meas. range begin 0.50
Meas. range end 5.00
Reference cell 10 mm
Calculation when measuring the absorbance value using the above NOVA photometers without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/l
[A sample * 1 / 0.2188]
1.000 4.57
Parameter list for programming the photometer SQ 118
01 Method V
02 Cat.No. -------
03 Measuring range 0.50 - 5.00
04 Units mg/l
05 Reaction time 0 0 min
06 Cell 10 mm
07 Wavelength 405 nm
08 Calibration Factor
09 Evaluation Linear
10 Factor 4.57
Calculation when measuring the absorbance value using the photometer SQ118 without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/l
[A sample * 4.57]
1.000 4.57
Materials
Related Links
Categories