Spectroquant® Prove Analytical Method: Vanadium in water
Intstruments and Reagents:
Spectroquant® VIS Spectrophotometer Prove 100 (Cat. No. 173016) or Spectroquant® UV/VIS Spectrophotometer Prove 300 (Cat. No. 173017) or Spectroquant® UV/VIS Spectrophotometer Prove 600 (Cat. No. 173018)
Potassium chloride for analysis (Cat. No. 104936)
Hydrochloric acid 1 mol/L (Cat. No. 109057)
Ethanol for analysis (Cat. No. 100983)
Water for analysis (Cat. No. 116754)
2-Thiophen-carboxylic hydrazide (Cat. No. T1388)
Membrane filter 0.45 μm pore diameter
Reagent 1:
Dissolve 6 g 2-thiophen-carboxylic hydrazide in 40 mL ethanol.
Reagent 2:
Place 12.5 mL potassium chloride solution, 0.2 mol/L and 3.25 mL hydrochloric acid, 0.2 mol/L in a 50-mL volumetric flask and make up to the mark with water.
Measurement range:
0.50 - 5.00 mg/L V at cell length 10 mm and 405 nm
Sample preparation:
The sample solution must be clear; if not, filter through a folded filter or membrane filter with 0.45 μm pore diameter.
Analysis:
Mix 10.0 mL of sample, 0.25 mL reagent 1, 0.30 mL reagent 2 in a test tube. Measure against water at 405 nm.
Parameter list for programming the photometer Spectroquant® Prove 100, Prove 300 and Prove 600
Method-designation Vanadium
Units mg/L
Citation V
Wavelength 405 nm
Cell 10 mm
Resolution 0.01
Meas. range 0.50 - 5.00
User range user defined
Zero point (E0) 0.000
calib. coefficient (A1) 4.57
Parameter list for programming the photometer Spectroquant® Pharo 100 and Pharo 300
Method number ----
Method-designation Vanadium
Wavelength 405 nm
Citation V
Units mg/L
Resolution 0.01
Zero point (E0) 0.000
Slope (E/C) 0.2188
Meas. range begin 0.50
Meas. range end 5.00
Reference cell 10 mm
Calculation when measuring the absorbance value using the above photometers without programming the method in the memory of the instrument:
Measured absorbance
of samples in A
Result in mg/L
[Asample * 4,57]
1.000 4.57
Materials
Related Links
Categories