Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8 PM CST on Friday evening, September 20th until 9 AM CST on Saturday morning, September 21st.
Please note that you will still have telephone and email access to our local offices during this period.
We apologize for any inconvenience this may cause.
Adsorbents for TLC and LPLC
We offer a comprehensive portfolio of adsorbents for the preparation of TLC plates as well as for LPLC (low pressure liquid Chromatography. All sorbents are standardized to ensure reliable analytical results.
Depending on the TLC plate you need to prepare, you can choose from a variety of sorbent materials, grades and particle sizes, which are available with or without binders and fluorescent indicators.
Features and benefits
  • Broad portfolio to suit different TLC and PLC applications
  • Standardized quality for reliable results
Loose Silica Sorbents for TLC and PLC plates (particle size 5–40 µm)
Product No. Description Sorbent / Coating Material*4 Comment Package Material Pack size / Content
1.07731.1000 Silica gel 60 G Silica gel 60 G Classical TLC plastic 1 kg
1.07731.5000 Silica gel 60 G Silica gel 60 G Classical TLC tin 5 kg
1.07731.9025 Silica gel 60 G Silica gel 60 G Classical TLC tin 25 kg
1.07730.1000 Silica gel 60 G F254 Silica gel 60 G F254*1 Classical TLC,
with Gypsum
plastic 1 kg
1.07730.5000 Silica gel 60 G F254 Silica gel 60 G F254*1 Classical TLC,
with Gypsum
tin 5 kg
1.07730.9025 Silica gel 60 G F254 Silica gel 60 G F254*1 Classical TLC,
with Gypsum
tin 25 kg
1.11678.1000 Silica gel 60 G F254 Silica gel 60 G F254*1 TLC plastic 1 kg
1.07736.1000 Silica gel 60 H Silica gel 60 H*3 TLC plastic 1 kg
1.07736.2500 Silica gel 60 H Silica gel 60 H*3 TLC tin 2.5 kg
1.07736.9025 Silica gel 60 H Silica gel 60 H*3 TLC tin 25 kg
1.11695.1000 Silica gel 60 H Silica gel 60 H*3 TLC plastic 1 kg
1.07739.1000 Silica gel 60 H F254 Silica gel 60 H *3F254*1 TLC plastic 1 kg
1.07739.2500 Silica gel 60 H F254 Silica gel 60 H *3F254*1 TLC tin 2.5 kg
1.07739.9025 Silica gel 60 H F254 Silica gel 60 H *3F254*1 TLC tin 25 kg
1.07741.1000 Silica gel 60 H F254+366 Silica gel 60 H *3F254+366*2 TLC plastic 1 kg
1.07747.1000 Silica gel 60 P F254 Silica gel 60 P F254*1 PLC, for
preparative work
plastic 1 kg
1.07747.2500 Silica gel 60 P F254 Silica gel 60 P F254*1 PLC, for
preparative work
tin 2.5 kg
1.07747.9025 Silica gel 60 PF254 Silica gel 60 P F254*1 PLC, for
preparative work
tin 25 kg
1.07748.1000 Silica gel 60 P F254+366 Silica gel 60 P F254+366*2 PLC, for
preparative work
plastic 1 kg
1.07748.2500 Silica gel 60 P F254 Silica gel 60 P F254*1 PLC, for
preparative work
tin 2.5 kg
1.07749.1000 Silica gel 60 P F254 Silica gel 60 P F254*1 PLC, for
preparative work
plastic 1 kg
1.07749.2500 Silica gel 60 PF254 Silica gel 60 P F254*1 PLC, for
preparative work
tin 2.5 kg
1.07749.9025 Silica gel 60 P F254 Silica gel 60 P F254*1 PLC, for
preparative work
tin 25 kg
288519-250G Silica gel 60 Silica gel 60 Particle size
5 - 25 µm
plastic 250g
Loose Aluminum Oxide Sorbents for TLC and PLC (particle size 5–40 µm)
Product No. Description Material Comment pH of 10% aqueous suspension Package Material Pack size/ Content
1.01090.2500 Aluminum oxide 60 G, neutral (type E) Aluminum oxide 60 G, neutral TLC, with Gypsum 7,5 Plastic 2.5 kg
1.01090.9025 Aluminum oxide 60 G, neutral (type E) Aluminum oxide 60 G, neutral TLC, with Gypsum 7,5 Plastic 25 kg
1.01092.0500 Aluminum oxide 60 G F254 neutral (type E) Aluminum oxide 60 G F254*1 neutral TLC, with Gypsum 7,5 Plastic 500 g
*1 F254 Fluorescent indicator
Other Sorbents for TLC
Product No. Description Particle Size Package Material Pack size/ Content
1.02330.0500 Cellulose microcrystalline < 20 µm plastic 500 g