Separation of Lipophilic Dye mixture on Silica gel 60
Separation of Lipophilic Dye mixture on Silica gel 60
Chromatographic Conditions
Plate TLC Silica gel 60 F254, 20x20cm
Article number 1. 05715.0001
Mobile Phase (v/v) toluene
Migration distance 10cm
Application volume 2 µL
Chamber normal chamber without chamber saturation
Detection Detection white light
Chromatographic Data
No. Compounds       Concentration (mg/ml) Solvent
1 to 7 p-Methoxyazobenzol
(52)
3.0 toluene
  Sudan II (79) 3.0 toluene
  Sudanviolett (89) 3.0 toluene
  Ceresviolett BRN (55) 1.0 toluene
Materials