Cellulose TLC and HPTLC Plates
Precise Analysis of Polar Substances
The organic sorbent cellulose is particularly suitable for the separation of hydrophilic substances by partition chromatography. We offer cellulose plates for demanding high-performance separations. Based on microcrystalline cellulose, the TLC plates are ideal for standard separations. Whereas the HPTLC plates use high-purity, rod-shaped microcrystalline cellulose, resulting in a highly reduced diffusion of analytes for demanding, high-performance separations.
We also provide special PEI cellulose plates (polyethylenimine-modified cellulose), which act as strong basic anion exchangers. This makes them the best choice for analyzing substances with exchange-active ionic groups, such as amino acids, peptides, nucleotides and nucleosides.
Cellulose plates are available with or without fluorescent indicators. They utilize a special fluorescent pigment that is stimulated to intense blue fluorescent emission by long-wave UV light of 366 nm and green fluoresent emission by short-wave UV light at 254 nm.
Typical applications of cellulose plates include the analysis of amino acids, carbohydrates, phosphates, nucleic acids and nucleic acid derivatives for:
  • Detection of significantly high increase of amino acids in clinical laboratories
  • 2-dimensional separations such as amino acid “fingerprints”
  • Metabolic studies
Cellulose Plates
Product No. Description Layer Sorbent/
Coating Material
Format/ cm Support/ Backing Pack size/ Content
1.05716.0001 TLC Cellulose Cellulose 20 x 20 glass 25 plates
1.05730.0001 TLC Cellulose Cellulose 10 x 20 glass 50 plates
1.05632.0001 TLC Cellulose Cellulose 10 x 10 glass 100 plates
1.05718.0001 TLC Cellulose F Cellulose F*1 20 x 20 glass 25 plates
1.05728.0001 TLC Cellulose F Cellulose F*1 10 x 20 glass 50 plates
1.05552.0001 TLC Cellulose Cellulose 20 x 20 aluminum 25 sheets
1.05563.0001 TLC Cellulose Cellulose 500 x 20 aluminum 1 roll
1.05574.0001 TLC Cellulose F Cellulose F*1 20 x 20 aluminum 25 sheets
1.05577.0001 TLC Cellulose Cellulose 20 x 20 plastic 25 sheets
1.05565.0001 TLC Cellulose F Cellulose F 20 x 20 plastic 25 sheets
1.05786.0001 HPTLC Cellulose HPTLC Cellulose 20 x 10 glass 50 plates
1.05787.0001 HPTLC Cellulose HPTLC Cellulose 10 x 10 glass 25 plates
1.15036.0001 HPTLC Cellulose F HPTLC Cellulose F*1 20 x 10 glass 50 plates
1.15035.0001 HPTLC Cellulose F HPTLC Cellulose F*1 10 x 10 glass 25 plates
1.16092.0001 HPTLC Cellulose HPTLC Cellulose 20 x 20 aluminum 25 sheets
1.05725.0001 TLC PEI Cellulose F PEI*2 Cellulose F*1 20 x 20 glass 25 plates
1.05579.0001 TLC PEI Cellulose F PEI*2 Cellulose F*1 20 x 20 plastic 25 sheets
Z122882-25EA HPTLC Cellulose PEI HPTLC Cellulose 20 x 20 plastic 25
*1 Fluorescent indicator wavelength 254/366 nm.
*2 PEI Cellulose plates should be stored cold (0-4°C) and dry to reduce deterioration. As plates become old they might get yellow colorations and should be discarded.