TLC analysis of Cholesterol and its Bile Acid Metabolites on HPTLC pre-coated plate RP-2 F 254s
TLC analysis of Cholesterol and its Bile Acid Metabolites on HPTLC pre-coated plate RP-2 F 254s
Conditions
plate HPTLC pre-coated plate RP-2 F 254s
eluent methanol/1N acetic acid 80/20 (v/v)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample application volume 300 nl
detection spray reagent: MnC12 - sulfuric acid heating up to 120 C for 5 min. UV 366 nm
Description
Application No. BOR180918
Featured Industry Life Science and Biopharma
Compound Class Lipikids