Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8 PM CST on Friday evening, September 20th until 9 AM CST on Saturday morning, September 21st.
Please note that you will still have telephone and email access to our local offices during this period.
We apologize for any inconvenience this may cause.
Kieselguhr and Mixed Layer Plates
Kieselguhr is a natural diatomaceous earth that can be used for the separation of polar or moderately polar compounds. For the mixed layer plates a combination of classical silica gel 60 and kieselguhr is utilized, providing favorable properties for certain applications such as separations of inorganic ions, herbicides or steroids.
Kieselguhr TLC Plates
Product No. Description Layer Sorbent/Coating Material Layer Thickness/ µm Format /cm Support/ Backing Pack Size / Content
1.05738.0001 TLC Kieselguhr F254 Kieselguhr F254*1 200 20 x 20 glass 25 plates
1.05568.0001 TLC Kieselguhr F254 Kieselguhr F254*1 200 20 x 20 aluminum 25 sheets
1.05567.0001 TLC Silica gel 60/Kieselguhr F254 Silica gel 60/
Kieselguhr F254*1
200 20 x 20 aluminum 25 sheets
*1 F254 Fluorescent indicator