Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8 PM CST on Friday evening, September 20th until 9 AM CST on Saturday morning, September 21st.
Please note that you will still have telephone and email access to our local offices during this period.
We apologize for any inconvenience this may cause.
Multiformat-Plates
Multiformat glass plates are pre-scored so that they can be easily snapped by hand into smaller sizes. The plates utilize the same proven silica gel coating as the corresponding non-scored TLC or HPTLC plate, delivering chromatograms of equally high quality.
The number of possible plates depends on the scoring. For example, for a 20 × 20 cm plate, scored in sections of 5 × 10, up to seven different formats are possible:
5 cm × 10 cm
5 cm × 20 cm
10 cm × 10 cm
10 cm × 15 cm
10 cm × 20 cm
15 cm × 20 cm
20 cm × 20 cm
Features and benefits
  • Simply snap to desired size
  • Same high quality as classical non-scored TLC and HPTLC plates
  • Up to 7 formats in one plate
How to Use Multiformat TLC and HPTLC Plates
Note: To prevent the glass backing from breaking in an uncontrolled and irregular manner, avoid putting plates directly on hot metal plates, drying cabinets or plate heaters after development and/or staining. When heat drying is necessary, please use distance holders of low thermal conductivity between the glass and the hot metal plate, i.e. glass rods or similar.
Materials
Product No. Description Layer Sorbent / Coating Material Plate size / cm Scored / cm Pack size/ Content number of plates possible
1.05620.0001 TLC Silica gel 60 F524 Multiformat pre-scored5 x 10 cm Multiformat silica gel 60 F254*1 20 x 20 5 x 10 25 plates 200
1.05608.0001 TLC Silica gel 60 F524Multiformat pre-scored 5 x 20 Multiformat silica gel 60 F254*1 20 x 20 5 x 20 20 plates 80
1.05635.0001 HPTLC Silica gel 60 Multiformat pre-scrored 5 x 5 cm  Multiformat silica gel 60 F254*1 10 x 10 5 x 5 25 plates 100
1.05644.0001 HPTLC Silica gel 60Multiformatpre-scrored 5 x 5 cm  Multiformat silica gel 60 10 x 10 5 x 5 100 plates 400
*1 F254 fluorescent indicator