[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Thin Layer Chromatography](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/thin-layer-chromatography)PLC Plates for Separations from Milligrams to Grams
# PLC Plates for Separations from Milligrams to Grams
Our Preparative layer (PLC) plates allow separation and purification of samples varying in quantity – from milligrams to grams. Samples are typically applied as a band across the width of the glass plate and analyzed by UV detection. To isolate the substance by extraction, it is simply scraped from the PLC plate. We offer PLC plates with both unmodified and modified layers, in a variety of thicknesses (0.5 mm to 2 mm), and with or without fluorescent indicators.
#### Features and benefits
- Separation and purification from milligram to gram quantities
- Thicker layers for high sample loading
- Same proven silica-binder technology as our analytical TLC plates
### PLC silica gel 60
Product No.DescriptionLayer Sorbent /Coating MaterialLayer Thickness/ mmFormat/ cmSupport/ BackingPack size/ Content
[1.13894.0001](https://www.sigmaaldrich.com/US/en/product/mm/113894)PLC Silica gel 60, 0.5 mmSilica gel 600,520 x 20glass20 plates
[1.05745.0001](https://www.sigmaaldrich.com/US/en/product/mm/105745)PLC silica gel 60, 2mmSilica gel 60220 x 20glass12 plates
[1.05744.0001](https://www.sigmaaldrich.com/US/en/product/mm/105744)PLC Silica gel 60 F254, 0.5 mmSilica gel 60 F254\*10,520 x 20glass20 plates
[1.13895.0001](https://www.sigmaaldrich.com/US/en/product/mm/113895)PLC Silica gel 60 F254, 1 mmSilica gel 60 F254\*1120 x 20glass15 plates
[1.05717.0001](https://www.sigmaaldrich.com/US/en/product/mm/105717)PLC Silica gel 60 F254, 2 mmSilica gel 60 F254\*1220 x 20glass12 plates
[1.05637.0001](https://www.sigmaaldrich.com/US/en/product/mm/105637)PLC Silica gel 60 F254+366, 2 mmSilica gel 60F F254 +366\*2220 x 20glass12 plates
[1.05434.0001](https://www.sigmaaldrich.com/US/en/product/mm/105434)PLC Silica gel 60 RP-18 F254s, 1 mmSilica RP-18 F254s\*3120 x 20glass15 plates
[Z265829-1PAK](https://www.sigmaaldrich.com/US/en/product/aldrich/z265829)PLC Silica gel 60 F254, 1 mmSilica gel 60 F254\*1120 x 20glass25 plates
[Z513059-1PAK](https://www.sigmaaldrich.com/US/en/product/aldrich/z513059)PLC Silica gel 60 F254, 2 mmSilica gel 60 F254\*1220 x 20glass25 plates
[Z513040-1PAK](https://www.sigmaaldrich.com/US/en/product/aldrich/z513040)PLC Silica gel 60 F254, 1.5 mmSilica gel 60 F254\*11,520 x 20glass25 plates
[Z513032-1PAK](https://www.sigmaaldrich.com/US/en/product/aldrich/z513032)PLC Silica gel 60 F254, 0.5 mmSilica gel 60 F254\*10,520 x 20glass25 plates
\*1 F254 Fluorescent indicator
\*2 F254+366 Fluorescent indicator
\*2 F254s Fluorescent indicator, acid stable
### PLC aluminum oxide 60 and 150
Product No.DescriptionLayer Sorbent/ Coating MaterialLayer Thickness/ mmFormat/ cmSupport/ BackingPack size/ Content
[1.05788.0001](https://www.sigmaaldrich.com/US/en/product/mm/105788)PLC Aluminum oxide 60 F254, 1..5 mmAluminum oxide 60 F254\*11,520 x 20glass12 plates
[1.05726.0001](https://www.sigmaaldrich.com/US/en/product/mm/105726)PLC Aluminum oxide 60 F254, 1..5 mmAluminum oxide 150 F254\*11,520 x 20glass12 plates
\*1 F254 Fluorescent indicator
__Related Articles__
- [Analysis of Catechins in Green Tea by TLC](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/analysis-of-catechins-in-green-tea-by-tlc)
- [High-Performance Thin-Layer Chromatography: A Fast and Efficient Fingerprint Analysis Method for Medicinal Plants](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/high-performance-thin-layer-chromatography)
- [High Performance Thin Layer Chromatography Plates](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/high-performance-thin-layer-chromatography-plates)
- [Identification of fixed oils by Thin layer chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/identification-of-fixed-oils-by-thin-layer-chromatography)
- [Identification of Glycyrrhizic Acid in Licorice](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/identification-of-glycyrrhizic-acid-in-licorice)
- [Tips & Tricks for Thin-Layer Chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tips-n-tricks-for-thin-layer-chromatography)
- [History of Thin-Layer Chromatography](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tlc-history)
- [Determination of Stevioside and Rebaudioside A in Stevia leaves, beverages and food additives using HPTLC-MS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/thin-layer-chromatography/tlc-ms-determination-of-stevioside-and-rebaudioside)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Thin Layer Chromatography (TLC)](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/tlc-plates-and-adsorbants)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.