Attention:
As we update our core systems, key features of the website including ordering and real-time price and availability, may be unavailable from 8 PM CST on Friday evening, September 20th until 9 AM CST on Saturday morning, September 21st.
Please note that you will still have telephone and email access to our local offices during this period.
We apologize for any inconvenience this may cause.
Premium Purity HPTLC Plates
Premium purity HPTLC plates are optimized for high-performance, completely contamination-free separations, such as demanding pharmacopoeia applications. These highly pure plates exhibit minimal background, even with medium-polar solvent systems.
Based on our proven 60 F254 HPTLC plate, premium purity plates are carefully wrapped in plastic-coated aluminum foil. This prevents deposition of plasticizers (such as phthalates), which could appear as unknown extra zones when using medium-polar solvent systems, such as toluene/ethyl acetate (95/5). Furthermore, it avoids staining by derivatization reagents, such as anis aldehyde. The separation performance of each premium purity plate is identical to the corresponding HPTLC plate.
Features and benefits
  • Optimized for high performance pharmacopeia applications
  • Completely contamination-free separations
  • Special wrapping prevents deposition of plasticizers
  • Minimal background even with medium-polar solvent systems
Premium Purity HPTLC Plates
Product No. Description Layer Sorbent/Coating Material Layer Thickness /µm Format/cm Support/Backing Pack size/Content
1.05648.0001 HPTLC silica gel 60 F254 premium purity HPTLC silica gel 60 F254 *1premium purity 200 20 x 10 glass 25 plates
*1 F254 Fluorescent indicator