Separation of Polycyclic Aromatic Hydrocarbons
The special HPTLC silica gel 60 RP-18 PAH concentrating zone plate is optimized for high-resolution separations of polycyclic aromatic hydrocarbons (PAH) according to DIN 38409-H13. These compounds result from pyrolysis or incomplete combustion of organic material. Main sources include the exhaust fumes of private and industrial furnaces, car exhaust, and tobacco smoke. Since some PAH are carcinogenic, maximum limits have been set for substances such as drinking water, and their determination is of great importance.
Product Table
Product No. Description Layer Sorbent/ Coating Material Layer thickness/ µm Format/ cm Support/ Backing Pack size/ Content
1.15037.0001 HPTLC Silica gel 60 RP-18 PAH concentrating zone 2.5 x 20 cm Silica gel 60 RP-18 PAH concentrating zone 2.5 x 20 cm 250 20 x 10 glass 25 plates
Categories