TLC Analysis of Stilbestrol Derivatives on HPTLC pre-coated plate RP-8 F 254s
TLC Analysis of Stilbestrol Derivatives on HPTLC pre-coated plate RP-8 F 254s
Conditions
plate HPTLC pre-coated plate RP-8 F 254s
eluent methanol/water 80/20 (v/v)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample application volume 200 nl
detection in-sJtu evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Application No. BOR180918A
Featured Industry Life Science and Biopharma
Compound Class Steroids