TLC Analysis of Nucleosides on HPTLC pre-coated plate RP-18 WF 254s
TLC Analysis of Sulfonamides on HPTLC pre-coated plate RP-18 WF 254s
Conditions
plate HPTLC pre-coated plate RP-18 WF 254s with concentrating zone (1.13124)
eluent Water/ethanol 95/5 (v/v)
migration distance 7 cm
chamber Normal chamber without chamber saturation
sample application volume 5 ul
detection In-situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
Description
Application No. BOR130918
Featured Industry Life Science and Biopharma
Compound Class Agriculture