TLC Analysis of a cosmetic active on HPTLC Silica gel 60 F254 (analyte set 2)
Conditions
sample preparation 1 g cosmetic samples was stirred with 10 ml 1-Propanol at room temperature and filtered with a 0.45 μm syringe filter. Application with CAMAG ATS4, 6 mm bandwise.
detector 254, 366 nm and white light (Derivatisation: Anisaldehyde- sulfuric acid)
eluent [A] petroleum benzine; [B] 1-Propanol ; (100:11, A:B)
chamber normal chamber without chamber saturation
migration distance 5 cm
plate 1.05628 HPTLC Silica gel 60 F254 10 cm x 10 cm (1.05628.0001)
Description
Featured Industry
Cosmetics, Personal Care, and Cleaning Products
suitability
application for TLC