TLC Analysis Of a Cosmetic Active on HPTLC Silica Gel 60 F254 (analyte set 1)
TLC Analysis Of a Cosmetic Active on HPTLC Silica Gel 60 F254 (analyte set 1)
Conditions
detector UV, 254 nm (Identification by coupling the TLC – MS Interface (CAMAG) with Microsaic 3500 MiD mass spectrometer (+)-ESI mode. Eluent: Acetonitrile / Water (95/5) + 0.1% TFA)
eluent [A] petroleum benzine; [B] acetone; (8:2, A:B)
chamber normal chamber without chamber saturation
migration distance 5 cm
plate 1.00934 HPTLC Silica gel 60 F254 MS-grade 20 cm x10 cm (1.00934.0001)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC